Learn More
Medchemexpress LLC Serotonin hydrochloride | 153-98-0 | 99.97% | 250 MG
SDP
Catalog No. 5000414488
Encompass_Preferred
Serotonin (5-Hydroxytryptamine) hydrochloride is a monoamine neurotransmitter found in the central nervous system (CNS). It acts as an endogenous 5-HT receptor agonist and can cross the blood-brain barrier. Additionally, it functions as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM, decreasing COMT reaction velocity and inducing robust hypersensitivity.
  • Monoamine neurotransmitter in the CNS.
  • Endogenous 5-HT receptor agonist.
  • Capable of crossing the blood-brain barrier.
  • Functions as a catechol O-methyltransferase (COMT) inhibitor.
  • Decreases COMT reaction velocity.
  • Induces robust hypersensitivity.
Catalog No. 50-004-14488 Supplier Medchemexpress LLC Supplier No. 250MGHYB1473
Add to Cart
Add to Cart
Serotonin (5-Hydroxytryptamine) hydrochloride is a monoamine neurotransmitter found in the central nervous system (CNS). It acts as an endogenous 5-HT receptor agonist and can cross the blood-brain barrier. Additionally, it functions as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM, decreasing COMT reaction velocity and inducing robust hypersensitivity.
  • Monoamine neurotransmitter in the CNS.
  • Endogenous 5-HT receptor agonist.
  • Capable of crossing the blood-brain barrier.
  • Functions as a catechol O-methyltransferase (COMT) inhibitor.
  • Decreases COMT reaction velocity.
  • Induces robust hypersensitivity.