Learn More
Abcam Serotonin hydrochloride (5-HT/5-Hydro x ytryptamine), 5-HT receptor agonist, 50MG
Supplier:  Abcam AB12052850MG
Encompass_Preferred
MW 212.67 Da, Purity >99%. Endogenous 5-HT receptor agonist. Neurotransmitter involved in diverse physiological functions such as mood, appetite, sleep, sex and temperature in addition to modulating cardiovascular function and the gastrointestinal system. Also a 5-HT transporter substrate.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7521