Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. S03705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03705G Supplier TCI America Supplier No. S03705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 153-98-0
Melting Point 169°C
Color Purple-White
Molecular Formula C10H13ClN2O
Quantity 5 g
MDL Number MFCD00012686
UN Number 2811
Synonym serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
InChI Key MDIGAZPGKJFIAH-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1O)C(=CN2)CCN.Cl
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
Molecular Weight (g/mol) 212.677
PubChem CID 160436
Formula Weight 212.68
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Serotonin Hydrochloride
Show More Show Less
EINECSNumber (5)-6504
RTECSNumber NM2571000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.