Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Serotonin Transporter Antibody (YA1638) | 100 UL
SDP

Catalog No. 5000440987
Encompass_Preferred

Serotonin Transporter Antibody (YA1638) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Serotonin Transporter. It is developed for research use, not for patient use. This antibody is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, and is shipped with blue ice. It is stored at -20°C for 1 year; avoid repeated freeze/thaw cycles.

  • Rabbit-derived, monoclonal IgG antibody.
  • Targets serotonin transporter.
  • Reacts with human, mouse, and rat samples.
  • Application: WB (Western Blot) with 1:500-1:1000 dilution.
  • Purity achieved via affinity chromatography.
  • Predicted and observed molecular weight: 70 kDa.
  • Endogenous sensitivity.

Catalog No. 50-004-40987 Supplier Medchemexpress LLC Supplier No. HYP81893100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serotonin Transporter Antibody (YA1638) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Serotonin Transporter. It is developed for research use, not for patient use. This antibody is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, and is shipped with blue ice. It is stored at -20°C for 1 year; avoid repeated freeze/thaw cycles.

  • Rabbit-derived, monoclonal IgG antibody.
  • Targets serotonin transporter.
  • Reacts with human, mouse, and rat samples.
  • Application: WB (Western Blot) with 1:500-1:1000 dilution.
  • Purity achieved via affinity chromatography.
  • Predicted and observed molecular weight: 70 kDa.
  • Endogenous sensitivity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.