Learn More
Cayman Chemical Serotonin (hydrochloride), Purity: ≥98%, CAS: 153-98-0, 100 mg
SDP
Catalog No. 502954999
Encompass_Preferred
A monoamine neurotransmitter produced in serotonergic neurons in the CNS and in enterochromaffin cells in the gastrointestinal tract that is important in the regulation of mood, sleep, vomiting, sexuality, and appetite
Catalog No. 50-295-4999 Supplier Cayman Chemical Supplier No. 14332100 MG
A monoamine neurotransmitter produced in serotonergic neurons in the CNS and in enterochromaffin cells in the gastrointestinal tract that is important in the regulation of mood, sleep, vomiting, sexuality, and appetite