Learn More
Medchemexpress LLC Serpin A3N Mouse P 50ug
SDP
Catalog No. 5000183497
Encompass_Preferred
Recombinant mouse Serpin A3N is a purified, His-tagged serine protease inhibitor expressed in Pichia pastoris for research applications. This reagent is intended for biochemical characterization, antibody development, and cell-based assays; it is supplied in a 50 μg pack with reported purity of ~92% and a molecular weight of ~46.8 kDa.
  • Suitable for biochemical assays and functional studies.
  • Useful for antibody generation and validation.
  • Expressed in Pichia pastoris for eukaryotic processing.
  • Includes N-terminal 6xHis tag for affinity purification or detection.
  • High reported purity (~92%) and defined molecular weight (~46.8 kDa).
  • Small pack size (50 μg) for pilot experiments and method development.
Catalog No. 50-001-83497 Supplier Medchemexpress LLC Supplier No. HYP7184150UG
Recombinant mouse Serpin A3N is a purified, His-tagged serine protease inhibitor expressed in Pichia pastoris for research applications. This reagent is intended for biochemical characterization, antibody development, and cell-based assays; it is supplied in a 50 μg pack with reported purity of ~92% and a molecular weight of ~46.8 kDa.
  • Suitable for biochemical assays and functional studies.
  • Useful for antibody generation and validation.
  • Expressed in Pichia pastoris for eukaryotic processing.
  • Includes N-terminal 6xHis tag for affinity purification or detection.
  • High reported purity (~92%) and defined molecular weight (~46.8 kDa).
  • Small pack size (50 μg) for pilot experiments and method development.