Learn More
Medchemexpress LLC Serpin A3N Mouse P 5ug
SDP
Catalog No. 5000183532
Encompass_Preferred
Recombinant mouse Serpin A3N protein produced in Pichia pastoris and purified with an N-terminal 6xHis tag. Supplied as a small-quantity aliquot suitable for biochemical assays, protease inhibition studies, and antibody validation in in vitro applications.
  • Recombinant mouse origin for species-specific studies.
  • Expressed in Pichia pastoris for eukaryotic expression characteristics.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Supplied in a 5 μg aliquot for low-volume assays and pilot experiments.
  • Suitable for protease inhibition assays, biochemical characterization, and antibody validation.
Catalog No. 50-001-83532 Supplier Medchemexpress LLC Supplier No. HYP718415UG
Recombinant mouse Serpin A3N protein produced in Pichia pastoris and purified with an N-terminal 6xHis tag. Supplied as a small-quantity aliquot suitable for biochemical assays, protease inhibition studies, and antibody validation in in vitro applications.
  • Recombinant mouse origin for species-specific studies.
  • Expressed in Pichia pastoris for eukaryotic expression characteristics.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Supplied in a 5 μg aliquot for low-volume assays and pilot experiments.
  • Suitable for protease inhibition assays, biochemical characterization, and antibody validation.