Learn More
Medchemexpress LLC Serpin A8/Angiotensi 10ug
SDP
Catalog No. 5000179985
Encompass_Preferred
Serpin A8 (or angiotensinogen) is a key regulator of the renin-angiotensin system affecting blood pressure fluid and electrolyte balance As a vasoconstrictor it directly affects vascular smooth muscle and affects myocardial contractility and heart rate through the sympathetic nervous system Serpin A8/Angiotensinogen Protein Rat (HEK293 His) is the recombinant rat-derived Serpin A8/Angiotensinogen protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79985 Supplier Medchemexpress LLC Supplier No. HYP7365110UG
Serpin A8 (or angiotensinogen) is a key regulator of the renin-angiotensin system affecting blood pressure fluid and electrolyte balance As a vasoconstrictor it directly affects vascular smooth muscle and affects myocardial contractility and heart rate through the sympathetic nervous system Serpin A8/Angiotensinogen Protein Rat (HEK293 His) is the recombinant rat-derived Serpin A8/Angiotensinogen protein expressed by HEK293 with C-His labeled tag