Learn More
Medchemexpress LLC Serpin A8/Angiotensi 500ug
SDP
Catalog No. 5000180619
Encompass_Preferred
Serpin A8 (or angiotensinogen) is an important regulator in the renin-angiotensin system fine-tuning blood pressure and electrolyte balance This multifunctional protein acts as a potent vasoconstrictor affecting vascular smooth muscle and affecting cardiac function via the sympathetic nervous system Serpin A8/Angiotensinogen Protein Mouse (HEK293 His) is the recombinant mouse-derived Serpin A8/Angiotensinogen protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80619 Supplier Medchemexpress LLC Supplier No. HYP73652500UG
Serpin A8 (or angiotensinogen) is an important regulator in the renin-angiotensin system fine-tuning blood pressure and electrolyte balance This multifunctional protein acts as a potent vasoconstrictor affecting vascular smooth muscle and affecting cardiac function via the sympathetic nervous system Serpin A8/Angiotensinogen Protein Mouse (HEK293 His) is the recombinant mouse-derived Serpin A8/Angiotensinogen protein expressed by HEK293 with C-His labeled tag