Learn More
Medchemexpress LLC Serpin A8/Angiotensi 50ug
SDP
Catalog No. 5000179979
Encompass_Preferred
Serpin A8/angiotensinogen protein is an important component of the renin-angiotensin system (RAS) which regulates blood pressure fluid balance and electrolyte homeostasis Serpin A8/Angiotensinogen Protein Human (HEK293 His) is the recombinant human-derived Serpin A8/Angiotensinogen protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79979 Supplier Medchemexpress LLC Supplier No. HYP7365350UG
Serpin A8/angiotensinogen protein is an important component of the renin-angiotensin system (RAS) which regulates blood pressure fluid balance and electrolyte homeostasis Serpin A8/Angiotensinogen Protein Human (HEK293 His) is the recombinant human-derived Serpin A8/Angiotensinogen protein expressed by HEK293 with C-His labeled tag