Learn More
Medchemexpress LLC Serpin H1 Human HE 500ug
SDP
Catalog No. 5000196626
Encompass_Preferred
Serpin H1 is a recombinant human SERPINH1 (Hsp47) protein produced in HEK293 cells and supplied lyophilized for in vitro research use. The protein is C-terminal His-tagged, functions as a collagen-specific molecular chaperone, and is provided with formulation and storage guidance for maintaining activity.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Contains a C-terminal His tag to facilitate detection and purification.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted mass 45.3 kDa; observed 48-70 kDa on SDS-PAGE due to glycosylation.
  • Lyophilized from PBS, pH 7.4, with optional 8% trehalose for stability.
  • Recommend reconstitution at ≥100 μg/mL and aliquot storage at -20°C or -80°C.
Catalog No. 50-001-96626 Supplier Medchemexpress LLC Supplier No. HYP71298500UG
Serpin H1 is a recombinant human SERPINH1 (Hsp47) protein produced in HEK293 cells and supplied lyophilized for in vitro research use. The protein is C-terminal His-tagged, functions as a collagen-specific molecular chaperone, and is provided with formulation and storage guidance for maintaining activity.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Contains a C-terminal His tag to facilitate detection and purification.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted mass 45.3 kDa; observed 48-70 kDa on SDS-PAGE due to glycosylation.
  • Lyophilized from PBS, pH 7.4, with optional 8% trehalose for stability.
  • Recommend reconstitution at ≥100 μg/mL and aliquot storage at -20°C or -80°C.