Learn More
Medchemexpress LLC Serpin H1 Human HE 5ug
SDP
Catalog No. 5000196830
Encompass_Preferred
Recombinant human Serpin H1 (heat shock protein 47) produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a lyophilized powder with >95% purity, the protein is suitable for research into collagen biosynthesis, protein folding, and chaperone function. Predicted molecular mass is 45.3 kDa; apparent molecular weight may be higher due to glycosylation.
  • Human-derived Serpin H1 expressed in HEK293 cells.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized format for convenient storage and transport.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Available in small quantities suitable for assay development.
  • Stable when aliquoted and stored at -20°C or -80°C.
Catalog No. 50-001-96830 Supplier Medchemexpress LLC Supplier No. HYP712985UG
Recombinant human Serpin H1 (heat shock protein 47) produced in HEK293 cells with a C-terminal 6xHis tag. Supplied as a lyophilized powder with >95% purity, the protein is suitable for research into collagen biosynthesis, protein folding, and chaperone function. Predicted molecular mass is 45.3 kDa; apparent molecular weight may be higher due to glycosylation.
  • Human-derived Serpin H1 expressed in HEK293 cells.
  • C-terminal 6xHis tag for affinity purification and detection.
  • Lyophilized format for convenient storage and transport.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Available in small quantities suitable for assay development.
  • Stable when aliquoted and stored at -20°C or -80°C.