Learn More
Aviva Systems Biology Corp SERPINA1 Antibody - C-terminal region (AVARP00015_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00015_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINA1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SERPINA1. Alternate names: PI, A1A, AAT, PI1, A1AT, PRO2275, alpha1ATApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-62450