Learn More
Aviva Systems Biology Corp SERPINA10 Antibody - middle region (ARP59235_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59235_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINA10. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SERPINA10. Alternate names: PZI, ZPIApplicationWB. Species reactivity: Human
Catalog No. 50-286-627