Learn More
ACROBiosystems Recombinant Protein;Human Serpin A1 Protein;HEK293;50UG;SE1,SerpinA1,Serpin A1,PI,A1A,AAT,PI1,A1AT,MGC9222,PRO2275,MGC23330,alpha1AT,SPAAT
Catalog No. 501658016
Encompass_Preferred
Recombinant Protein;Human Serpin A1 Protein;50UG;Human Serpin A1, His Tag (SE1-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 418 (Accession # AAH11991).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8016 Supplier ACROBiosystems Supplier No. SE1H522650UG
Recombinant Protein;Human Serpin A1 Protein;50UG;Human Serpin A1, His Tag (SE1-H5226) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 418 (Accession # AAH11991).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.