Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 12MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: AACT/1452. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HeLa Cells. Tonsil, Pancreas or Histiocytoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Antichymotrypsin (AACT) protein fragment (aa49-187). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 65-76kDa.Chromosome: 14q32.1. Gene Symbol: SERPINA3. Gene ID: 12. Ensemble: ENSG00000196136. Uniprot: NP_001076.
Catalog No. 50-188-4623