Learn More
Aviva Systems Biology Corp SERPINA3 Antibody - middle region (ARP51880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SERPINA3. Alternate names: AACT, ACT, GIG24, GIG25, MGC88254ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-60636