Learn More
ACROBiosystems Recombinant Protein;Human Serpin A3 Protein;HEK293;1MG;SE3,SERPINA3,Serpin A3,ACT,AACT,GIG24,GIG25
Supplier:  ACROBiosystems SE3H52211MG
Encompass_Preferred
Recombinant Protein;Human Serpin A3 Protein;1MG;Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession # AAH03559).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.6 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8369