Learn More
Aviva Systems Biology Corp SERPINA5 Antibody - C-terminal region (ARP53555_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53555_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SERPINA5. Alternate names: PAI3, PCI, PLANH3, PROCI, PAI-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61045