Learn More
Biomatik Corporation Serpinb14 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10355
Encompass_Preferred
Serpinb14 Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to SERPINB14. The Serpinb14 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Chicken Ovalbumin (Native Protein). Applications: ELISA, WB. Target Synonyms: SERPINB14. Accession Number: P01012. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6756