Learn More
Aviva Systems Biology Corp SERPINB6 Antibody - middle region (ARP59249_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINB6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SERPINB6. Alternate names: CAP, DKFZp686I04222, MGC111370, MSTP057, PI6, PTI, SPI3, DFNB91ApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-286-631