Learn More
Aviva Systems Biology Corp SERPINB7 Antibody - N-terminal region (ARP59251_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINB7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SERPINB7. Alternate names: DKFZp686D06190, MEGSIN, MGC120014, MGC120015ApplicationWB. Species reactivity: Human
Catalog No. 50-286-632