Learn More
Aviva Systems Biology Corp SERPINB8 Antibody - middle region (ARP57800_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57800_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINB8. It was validated on Western Blot. SERPINB8,serpin peptidase inhibitor, clade B (ovalbumin), member 8,PI8; CAP2; SERPINB8,serpin B8,serpin B8,PI-8,OTTHUMP00000067002,OTTHUMP00000180653,peptidase inhibitor 8,cytoplasmic antiproteinase 2,protease inhibitor 8 (ovalbumin type),serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 8The immunogen is a synthetic peptide directed towards the middle region of human SERPINB8. Alternate names: CAP2, PI8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-286-383