Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Serpin D1 Protein, His Tag, 1 mg

Catalog No. 501658502
Encompass_Preferred

Human Serpin D1, also known as Heparin cofactor II (HC-II), is a member of the serpin family expressed primarily in the liver. This thrombin inhibitor is activated by glycosaminoglycans such as heparin or dermatan sulfate. The protein is expressed in human HEK293 cells, contains amino acids Gly 20 to Ser 499, and features a polyhistidine tag at the C-terminus.

  • Purity: ≥90% as determined by SDS-PAGE
  • Calculated molecular weight: 55.7 kDa
  • Observed migration: 66-80 kDa under reducing conditions due to glycosylation
  • Endotoxin level: <1.0 EU per μg by LAL or rFC method
  • Formulation: Lyophilized from 0.22 μm filtered 50 mM Tris, 150 mM NaCl, pH 7.5 with trehalose

Catalog No. 50-165-8502 Supplier ACROBiosystems Supplier No. HC2H52241MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Serpin D1, also known as Heparin cofactor II (HC-II), is a member of the serpin family expressed primarily in the liver. This thrombin inhibitor is activated by glycosaminoglycans such as heparin or dermatan sulfate. The protein is expressed in human HEK293 cells, contains amino acids Gly 20 to Ser 499, and features a polyhistidine tag at the C-terminus.

  • Purity: ≥90% as determined by SDS-PAGE
  • Calculated molecular weight: 55.7 kDa
  • Observed migration: 66-80 kDa under reducing conditions due to glycosylation
  • Endotoxin level: <1.0 EU per μg by LAL or rFC method
  • Formulation: Lyophilized from 0.22 μm filtered 50 mM Tris, 150 mM NaCl, pH 7.5 with trehalose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.