Learn More
ACROBiosystems Recombinant Protein;Human Serpin D1 / Heparin Cofactor II Protein;HEK293;50UG;HC2,SerpinD1,HCF2,Serpin D1,HLS2,HC-II,HCF-2
Supplier:  ACROBiosystems HC2H522450UG
Encompass_Preferred
Recombinant Protein;Human Serpin D1 / Heparin Cofactor II Protein;50UG;Human Serpin D1, His Tag (HC2-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Ser 499 (Accession # AAH35028).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.7 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8501