Learn More
Aviva Systems Biology Corp SERPINE1 Antibody - C-terminal region (ARP47470_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47470_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SERPINE1. Alternate names: PAI, PAI-1, PAI1, PLANH1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59559