Learn More
Aviva Systems Biology Corp SERPINE1 Antibody - middle region (ARP59233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINE1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SERPINE1. Alternate names: PAI, PAI-1, PAI1, PLANH1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-286-626