Learn More
Aviva Systems Biology Corp SERPINF1 Antibody - N-terminal region (AVARP20020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP20020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SERPINF1. Alternate names: OI6, OI12, PEDF, EPC-1, PIG35ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-62651