Learn More
ACROBiosystems ACROBiosystems Human Serpin F1 / PEDF Protein, His Tag
Catalog No. 501956988
Encompass_Preferred
Recombinant Protein;100UG;Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Pro 418 (Accession # NP_002606).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-6988 Supplier ACROBiosystems Supplier No. SE1H5221100UG
Recombinant Protein;100UG;Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Pro 418 (Accession # NP_002606).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).