Learn More
ACROBiosystems Recombinant Protein;Human Serpin F1 / PEDF Protein;HEK293;1MG;SE1,SERPINF1,Serpin F1,PEDF,PIG35,EPC-1,SERPINF-1,EPC1,PIG-35
Catalog No. 501658296
Encompass_Preferred
Recombinant Protein;Human Serpin F1 / PEDF Protein;1MG;Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Pro 418 (Accession # NP_002606).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8296 Supplier ACROBiosystems Supplier No. SE1H52211MG
Add to Cart
Add to Cart
Recombinant Protein;Human Serpin F1 / PEDF Protein;1MG;Human Serpin F1, His Tag (SE1-H5221) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Pro 418 (Accession # NP_002606).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE).