Learn More
Aviva Systems Biology Corp SERPINF2 Antibody - N-terminal region (ARP59189_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59189_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SERPINF2. Alternate names: A2AP, AAP, ALPHA-2-PI, API, PLIApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-286-613