Learn More
ACROBiosystems Recombinant Protein;Human Serpin H1 Protein;HEK293;1MG;SE1,SERPINH1,Serpin H1,HSP47,HSP-47,CBP1,CBP2,SERPINH2,PIG14,AsTP3,Colligin,OI10,PPROM,RA-A47,gp46
Catalog No. 501658307
Encompass_Preferred
Recombinant Protein;Human Serpin H1 Protein;1MG;Human Serpin H1, His Tag (SE1-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 418 (Accession # NP_001226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.3 kDa. The protein migrates as 47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8307 Supplier ACROBiosystems Supplier No. SE1H52241MG
Recombinant Protein;Human Serpin H1 Protein;1MG;Human Serpin H1, His Tag (SE1-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Leu 418 (Accession # NP_001226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.3 kDa. The protein migrates as 47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.