Learn More
Aviva Systems Biology Corp SERPINI2 Antibody - middle region (ARP56686_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56686_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SERPINI2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SERPINI2. Alternate names: MEPI, PANCPIN, PI14, TSA2004ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-194