Learn More
Fisherbrand™ Serrated Hemostat
Catalog No. 12000171 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
12-000-171 Stainless Steel
1 options
Not for internal use
Catalog No. 12-000-171
Add to Cart
Edge
Add to Cart
Not for internal use
For clamping small blood vessels.
  • Locking mechanism allows clamp to hold vessel securely
  • Features serrated jaws for grip
  • Replacement tool for use with Fisher Scientific Dissection Kit with Black Zipper Case
Specifications
Product Type Serrated Hemostat
For Use With (Equipment) Fisher Scientific Dissection Kit
Material Stainless Steel
Jaw Type 25° Curved Jaws
Model PL-77