Learn More
Sigma Aldrich Fine Chemicals Biosciences Serrated natural rubber se
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5127101PAK
Encompass
Closure has a turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on a hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck making each serration a suction sealing point.Note for extra security when using large bore transfer needles fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings on next page.
Catalog No. 50-290-5670