Learn More
Sigma Aldrich Fine Chemicals Biosciences Serrated silicone rubber s
Catalog No. 502891561
Encompass
Translucent silicone rubber septa. Contain no pigments, waxes, or molding agents. Good for high heat applications (200C continuous or 400C intermittent). Autoclavable for sterile applications. Excellent resistance to deterioration due to sunlight and ozone
Catalog No. 50-289-1561 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5128691PAK
Translucent silicone rubber septa. Contain no pigments, waxes, or molding agents. Good for high heat applications (200C continuous or 400C intermittent). Autoclavable for sterile applications. Excellent resistance to deterioration due to sunlight and ozone