Learn More
ATCC Burkholderia cepacia; 6 x 0.3mL
Catalog No. 502380497
Encompass
ATCC 25416-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 25416™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-0497 Supplier ATCC Supplier No. 25416MINIPACK
ATCC 25416-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 25416™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.