Learn More
ATCC Serratia marcescens
Supplier:  ATCC 14756
Encompass
Serratia marcescens strain PCI 1107 is a whole genome-sequenced strain that was isolated from Fort Detrick. This bacterium has applications in testing membrane filters, antimicrobial agents, and antimicrobial handwashing formulations.
Catalog No. 50-238-3257