Learn More
AdipoGen Serratol (5 mg)
SDP
Supplier:  AdipoGen AGCN20483M005
Encompass_Preferred
Chemical. CAS: 67814-27-1. Formula: C20H34O. MW: 290.5. Isolated from Boswellia serrata. Component of the frankincense essential oil. Potent activator of transient receptor potential vanilloid 3 (TRPV3), a heat-sensitive ion channel involved in skin heat sensitivity, thermoregulation and neurological activities. More powerful TRPV3 agonist than incensol (Prod. No. AG-CN2-0482) and incensol acetate (Prod. No. AG-CN2-0484). Anti-protozoal agent.
Catalog No. 50-205-3666