Learn More
Fine Science Tools SERREFINES/BULLDOG/CURVED 35MM
SDP
Catalog No. NC9646886
Encompass
Product: Bulldog Serrefine - Curved/35mm/ Shape: Curved/ Jaw Dimensions: 10.5 x 2.3mm/ Clamping Pressure: Medium/ Alloy: Stainless Steel/ Length: 35mm/ Serrated: Yes/ Unit: Each
Catalog No. NC9646886 Supplier Fine Science Tools Supplier No. 1805135
Product: Bulldog Serrefine - Curved/35mm/ Shape: Curved/ Jaw Dimensions: 10.5 x 2.3mm/ Clamping Pressure: Medium/ Alloy: Stainless Steel/ Length: 35mm/ Serrated: Yes/ Unit: Each