Learn More
Medchemexpress LLC Sertaconazole nitrate | 99592-39-9 | 99.2% | 100 MG
SDP
Catalog No. 5000408191
Encompass_Preferred
Sertaconazole nitrate is a broad-spectrum topical antifungal agent that exhibits anti-inflammatory activity by activating a p38-COX-2-PGE2 pathway. It also acts as a microtubule inhibitor, demonstrating an antiproliferative effect, inducing apoptosis and autophagy, and inhibiting cell migration. This compound is widely used in research for its diverse biological activities.
  • Inhibits 150 strains of yeasts, including six Candida species
  • Exhibits anti-inflammatory activity by activating p38-COX-2-PGE2 pathway
  • Induces strong mitotic arrest and antiproliferative effects
  • Induces apoptosis through the p53 pathway
  • Inhibits cell migration
  • Induces autophagy in A549 and H460 cells
  • Suppresses TPA-induced ear edema in vivo
Catalog No. 50-004-08191 Supplier Medchemexpress LLC Supplier No. HYB0736A100MG
Sertaconazole nitrate is a broad-spectrum topical antifungal agent that exhibits anti-inflammatory activity by activating a p38-COX-2-PGE2 pathway. It also acts as a microtubule inhibitor, demonstrating an antiproliferative effect, inducing apoptosis and autophagy, and inhibiting cell migration. This compound is widely used in research for its diverse biological activities.
  • Inhibits 150 strains of yeasts, including six Candida species
  • Exhibits anti-inflammatory activity by activating p38-COX-2-PGE2 pathway
  • Induces strong mitotic arrest and antiproliferative effects
  • Induces apoptosis through the p53 pathway
  • Inhibits cell migration
  • Induces autophagy in A549 and H460 cells
  • Suppresses TPA-induced ear edema in vivo