Learn More
Medchemexpress LLC Sertaconazole nitrate | 99592-39-9 | 99.2% | 25 MG
SDP
Catalog No. 5000450965
Encompass_Preferred
Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent that exhibits anti-inflammatory activity by activating a p38-COX-2-PGE2 pathway. It is also a microtubule inhibitor, demonstrating antiproliferative effects, inducing apoptosis and autophagy, and inhibiting cell migration.
  • Broad-spectrum topical antifungal agent
  • Exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway
  • Microtubule inhibitor
  • Shows antiproliferative effect
  • Induces apoptosis and autophagy
  • Can inhibit the migration of cells
Catalog No. 50-004-50965 Supplier Medchemexpress LLC Supplier No. 25MGHYB0736A
Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent that exhibits anti-inflammatory activity by activating a p38-COX-2-PGE2 pathway. It is also a microtubule inhibitor, demonstrating antiproliferative effects, inducing apoptosis and autophagy, and inhibiting cell migration.
  • Broad-spectrum topical antifungal agent
  • Exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway
  • Microtubule inhibitor
  • Shows antiproliferative effect
  • Induces apoptosis and autophagy
  • Can inhibit the migration of cells