Learn More
Medchemexpress LLC Sertaconazole nitrate | 99592-39-9 | 99.2% | 500.78 | 10 MG
SDP
Catalog No. 5000365514
Encompass_Preferred
Sertaconazole nitrate is a broad-spectrum topical antifungal agent that exhibits anti-inflammatory activity by activating a p38-COX-2-PGE2 pathway. It also acts as a microtubule inhibitor, demonstrating antiproliferative effects, inducing apoptosis and autophagy, and inhibiting cell migration.
  • Broad-spectrum antifungal activity against 150 yeast strains, including six Candida species (mean MIC of 0.77 μg/mL in vitro).
  • Activates p38 MAP kinase and increases PGE2 release via COX-2 in keratinocytes.
  • Induces mitotic arrest by depolymerizing microtubules, leading to antiproliferation.
  • Induces apoptosis through the p53 pathway and autophagy in various cell types.
  • Inhibits migration of HeLa cells in a concentration-dependent manner.
  • Suppresses TPA-induced ear edema in CD-1 mice, reducing inflammation.
Catalog No. 50-003-65514 Supplier Medchemexpress LLC Supplier No. 10MGHYB0736A
Sertaconazole nitrate is a broad-spectrum topical antifungal agent that exhibits anti-inflammatory activity by activating a p38-COX-2-PGE2 pathway. It also acts as a microtubule inhibitor, demonstrating antiproliferative effects, inducing apoptosis and autophagy, and inhibiting cell migration.
  • Broad-spectrum antifungal activity against 150 yeast strains, including six Candida species (mean MIC of 0.77 μg/mL in vitro).
  • Activates p38 MAP kinase and increases PGE2 release via COX-2 in keratinocytes.
  • Induces mitotic arrest by depolymerizing microtubules, leading to antiproliferation.
  • Induces apoptosis through the p53 pathway and autophagy in various cell types.
  • Inhibits migration of HeLa cells in a concentration-dependent manner.
  • Suppresses TPA-induced ear edema in CD-1 mice, reducing inflammation.