Learn More
Medchemexpress LLC Sertindole-d4 | 1794737-42-0 | 98.6% | C24H22D4ClFN4O | 1 MG
SDP
Catalog No. 5000355286
Encompass_Preferred
Sertindole-d4 is a deuterium-labeled form of Sertindole, a well-known neuroleptic medication. This compound is primarily used in research applications, offering stable isotopic labeling for various analytical techniques. It is particularly valuable as a tracer for quantitation during drug development and can serve as an internal standard for precise quantitative analysis using methods like NMR, GC-MS, or LC-MS. Deuterium substitution can also influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound for research.
  • Acts as a tracer in drug development processes.
  • Suitable as an internal standard for quantitative analysis.
  • Potential for studying pharmacokinetic and metabolic profiles.
  • Useful in NMR, GC-MS, and LC-MS applications.
Catalog No. 50-003-55286 Supplier Medchemexpress LLC Supplier No. HY14543S1MG
Sertindole-d4 is a deuterium-labeled form of Sertindole, a well-known neuroleptic medication. This compound is primarily used in research applications, offering stable isotopic labeling for various analytical techniques. It is particularly valuable as a tracer for quantitation during drug development and can serve as an internal standard for precise quantitative analysis using methods like NMR, GC-MS, or LC-MS. Deuterium substitution can also influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound for research.
  • Acts as a tracer in drug development processes.
  • Suitable as an internal standard for quantitative analysis.
  • Potential for studying pharmacokinetic and metabolic profiles.
  • Useful in NMR, GC-MS, and LC-MS applications.