Learn More
Medchemexpress LLC Sertindole-d4 | 1794737-42-0 | 98.6% | C24H22D4ClFN4O | 10MG
SDP
Catalog No. 5000261919
Encompass_Preferred
Sertindole-d4 is the deuterium-labeled analogue of sertindole supplied as a stable-isotope internal standard for analytical workflows. Provided as a sealed powder with high purity and defined storage conditions, it supports accurate quantitation, ADME studies, and metabolite identification by LC-MS.
  • Deuterium-labeled analogue for use as an internal standard.
  • High purity suitable for analytical applications.
  • Suitable for LC-MS quantitation and metabolite identification.
  • Provided as a sealed powder for stable storage.
  • Specific storage recommendations for long-term stability.
Catalog No. 50-002-61919 Supplier Medchemexpress LLC Supplier No. HY14543S10MG
Sertindole-d4 is the deuterium-labeled analogue of sertindole supplied as a stable-isotope internal standard for analytical workflows. Provided as a sealed powder with high purity and defined storage conditions, it supports accurate quantitation, ADME studies, and metabolite identification by LC-MS.
  • Deuterium-labeled analogue for use as an internal standard.
  • High purity suitable for analytical applications.
  • Suitable for LC-MS quantitation and metabolite identification.
  • Provided as a sealed powder for stable storage.
  • Specific storage recommendations for long-term stability.