Learn More
Medchemexpress LLC Sertindole-d4 | 1794737-42-0 | MFCD25977368 | 98.6% | 444.97 | C24H22D4ClFN4O | 5 MG
SDP
Catalog No. 5000221315
Encompass_Preferred
Sertindole-d4 is the deuterium-labeled analog of the antipsychotic sertindole, supplied as an isotope-labeled reference standard for analytical and research applications. Intended for research use only, it is commonly used as an internal standard in mass spectrometry, tracer studies, and quantitative bioanalysis due to defined isotopic labeling and high purity.
  • Labeled with deuterium for stable isotope reference.
  • High purity suitable for analytical and quantitative work.
  • Consistent isotopic composition for reliable mass spectrometry quantitation.
  • Provided in accurately weighed small quantities for trace-level studies.
  • Accompanied by a product datasheet and certificate of analysis.
Catalog No. 50-002-21315 Supplier Medchemexpress LLC Supplier No. HY14543S5MG
Sertindole-d4 is the deuterium-labeled analog of the antipsychotic sertindole, supplied as an isotope-labeled reference standard for analytical and research applications. Intended for research use only, it is commonly used as an internal standard in mass spectrometry, tracer studies, and quantitative bioanalysis due to defined isotopic labeling and high purity.
  • Labeled with deuterium for stable isotope reference.
  • High purity suitable for analytical and quantitative work.
  • Consistent isotopic composition for reliable mass spectrometry quantitation.
  • Provided in accurately weighed small quantities for trace-level studies.
  • Accompanied by a product datasheet and certificate of analysis.