Learn More
Selleck Chemical LLC Sertraline HCl S4053-10mg
SDP
Catalog No. 503108124
Encompass_Preferred
Sertraline HCl (CP-51974-1) is a 5-HT antagonist with Ki of 13 nM
Catalog No. 50-310-8124 Supplier Selleck Chemical LLC Supplier No. S405310MG
Sertraline HCl (CP-51974-1) is a 5-HT antagonist with Ki of 13 nM