Learn More
TARGETMOL CHEMICALS INC Sertraline hydrochloride 25MG
SDP
Catalog No. 502880569
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Sertraline hydrochloride (Sertraline HCl) is a selective serotonin uptake inhibitor that is used in the treatment of depression. Purity 99.86%
Catalog No. 50-288-0569 Supplier TARGETMOL CHEMICALS INC Supplier No. T048225MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Sertraline hydrochloride (Sertraline HCl) is a selective serotonin uptake inhibitor that is used in the treatment of depression. Purity 99.86%