Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sertraline hydrochloride

Catalog No. p-7139376
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462190010 1 g
AC462190050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462190010 Supplier Thermo Scientific Chemicals Supplier No. 462190010
Only null left
Add to Cart

CAS: 79559-97-0 | C17H17Cl2N·HCl | 342.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79559-97-0
Molecular Formula C17H17Cl2N·HCl
Molecular Weight (g/mol) 342.69
InChI Key BLFQGGGGFNSJKA-XHXSRVRCSA-N
PubChem CID 63009
ChEBI CHEBI:9124

Specifications

Melting Point 245°C
Quantity 1 g
Merck Index 15,8606
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 342.69
Chemical Name or Material Sertraline hydrochloride
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.