Learn More
Medchemexpress LLC Sertraline hydrochloride | 79559-97-0 | >99.9% | 342.69 | 10 MG
SDP
Catalog No. 5000365625
Encompass_Preferred
Sertraline hydrochloride is an antidepressant classified as a selective serotonin reuptake inhibitor (SSRI). It is researched for various conditions such as major depressive disorder and obsessive-compulsive disorder.
  • Functions as a selective serotonin reuptake inhibitor (SSRI) antidepressant.
  • Researched for major depressive disorder and obsessive-compulsive disorder.
  • Inhibits high mobility group box 1 (HMGB1)- or tumor necrosis factor-α (TNF-α)-induced microglial activation.
  • Produces optimal anti-depression effects in chronic unpredictable mild stress (CUMS)-exposed mice.
  • Significantly reduces immobility time during forced swim test in female animals.
  • Does not alter spontaneous locomotor activity in all three genotypes regardless of sex difference.
Catalog No. 50-003-65625 Supplier Medchemexpress LLC Supplier No. 10MGHYB0176A
Sertraline hydrochloride is an antidepressant classified as a selective serotonin reuptake inhibitor (SSRI). It is researched for various conditions such as major depressive disorder and obsessive-compulsive disorder.
  • Functions as a selective serotonin reuptake inhibitor (SSRI) antidepressant.
  • Researched for major depressive disorder and obsessive-compulsive disorder.
  • Inhibits high mobility group box 1 (HMGB1)- or tumor necrosis factor-α (TNF-α)-induced microglial activation.
  • Produces optimal anti-depression effects in chronic unpredictable mild stress (CUMS)-exposed mice.
  • Significantly reduces immobility time during forced swim test in female animals.
  • Does not alter spontaneous locomotor activity in all three genotypes regardless of sex difference.